Labels for LABXPERT™ printer, type B-492, Freezerbondz™
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BRDY105951EA 148.35 EUR
BRDY105951 BRDY105953 BRDY105944 BRDY105946 BRDY105948 BRDY105949 BRWWX-120-492
Labels for LABXPERT™ printer, type B-492, Freezerbondz™
FreezerBondz™ material is a low-profile label used on frozen surfaces or on room-temperature surfaces that will eventually be stored in liquid nitrogen (–196 °C). The improved B-492 version FreezerBondz features enhanced performance and related changes to the label size.
  • Identify frosted, frozen or room temperature vials (with overlap)
  • Superior solvent resistance
  • Opaque enough to cover legally protected patient information for compliance with HIPAA requirements