Netherlands
Order Entry
Column heaters, Sidewinder™
Column heaters, Sidewinder™
Supplier:  Restek
Certificates
undefined
Column heaters, Sidewinder™
Supplier:  Restek
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • Temperature control module and column holder, 5 - 85 °C, ±1 °C, 100 mm length
    Temperature control module and column holder, 5 - 85 °C, ±1 °C, 100 mm length
    REST26517
    26517
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Supplier Article Number: 26517
    Local Article Number: REST26517
    Specifications:
    Specifications
    Environmentally Preferable
  • Temperature control module and column holder, 5 - 85 °C, ±1 °C, 250 mm length
    Temperature control module and column holder, 5 - 85 °C, ±1 °C, 250 mm length
    REST26516
    26516
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Supplier Article Number: 26516
    Local Article Number: REST26516
    Specifications:
    Specifications
    Environmentally Preferable
  • Heater/Cooler temperature control module and column holder, 5 - 55 °C, ±0,2 °C, 300 mm length
    Heater/Cooler temperature control module and column holder, 5 - 55 °C, ±0,2 °C, 300 mm length
    REST26518
    26518
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Supplier Article Number: 26518
    Local Article Number: REST26518
    Specifications:
    Specifications
    Environmentally Preferable