Face shield, F800
Leverancier: Scott Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SCTS2014617EA 22.34 EUR
SCTS2014617
Face shield, F800
F800 face shield made from translucent PC consists of a visor frame and the browguard. It has high levels of protection with a wide field of vision.
  • Anti-fog option
  • Built in chinguard provides additional protection
  • Central face shield attachment minimises visor replacement cost
This browguard has a new ratchet headgear, with positive lock positions across a wide range of head sizes and a comfortable shaped headgear to maximise wearer comfort.
Certificaten: EN 166 B39