Powered air respirator (PAPR) system, DURAFLOW
Leverancier: Scott Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DURAFLOW
SCTS2031662EA 807.15 EUR
SCTS2031662
Powered air respirator (PAPR) system, DURAFLOW
Ademhalingsmaskers Ademhalingssystemen
The DURAFLOW powered air respirator is a durable unit providing electronic flow control, in-use status monitoring and filter and battery alarms in a durable blower designed to meet the demands of the most aggressive working environments.
  • Lightweight, low profile ergonomic design provides comfort even over extended durations of hard work
  • Intelligence unit to monitor the flow rate in real time
  • Alarm sounds for incorrect flow rate
This respirator system features a durable body to withstand arduous environments and a range of design features including real-time air flow control, audible/visual alarms, two standard duration lithium-ion batteries and LED operation displays.