Filters for Profile and Promask face masks
Leverancier: Scott Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SCTS2032213EA 228.2 EUR
SCTS2032213
Filters for Profile and Promask face masks
These Pro2 filters have a safe bayonet fit locking system and unique covers to provide protection from splashes, sparks and contaminants.
  • Unique protective cover with rear facing grill inlet
  • Low breathing resistance for efficient and comfortable operation
  • Low profile and unobtrusive
  • Provide excellent balance on the facepiece
  • Microfibre paper media is used instead of any electrostatic filtering methods
Certificaten: EN 143; EN 14387