Algicide for water baths, Aquastab
Leverancier: Lauda
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LAUDLZB129EA 137 EUR
LAUDLZB129 LAUDLZB429 LAUDLZB929
Algicide for water baths, Aquastab
Reinigingsmiddelen Desinfectiemiddelen
Effective algicide, only to be used with water in bath thermostats.
  • Reduces contamination of water by bacteria and fungi
  • Use 5 ml of solution per 10 litres of water
Temperature range: 5 to 90 °C
Density: 0,95 to 1,05 g/ml
Colour: blue
Opgelet: Use biocides safely. Always read the label and product information before use.