Streptavidin-sepharose beads
Leverancier: BioVision
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIOV6565-2EA 209 EUR
BIOV6565-2 BIOV6565-5 BIOV6565-10
Streptavidin-sepharose beads
Sepharose beads conjugated to streptavidin for binding to free biotin or biotin based products.