Horse serum
Leverancier: Biowest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S0910-500EA 77.2 EUR
S0910-500 S091H-100 S0910-100 S091H-500
Horse serum
Serum
Horse serum asceptically collected from healthy animals. Used as a blocking agent or as a control for immunoassays.