Semi-automatic Kjeldahl distillation unit, UDK 139
Leverancier: VELP Scientifica
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
701-0226EA 4344 EUR
VELPF30200130
Semi-automatic Kjeldahl distillation unit, UDK 139
The UDK 139 is a semi-automatic model offering greater automation and a wider range of programming options compared to the UDK 129. This unit runs automatically, after setting distillation time, sodium hydroxide and water addition and steam generation output level between 10 and 100%. The high precision pumps ensure constant accurate dosing of reagents.
  • Automatic NaOH and H₂O addition
  • Delay time (Devarda alloy analysis) and selectable distillation time
  • Distillation residues removal
  • Language selection
  • Safety lever and sensors and reagent level warning to protect the user
All the UDK series distillation units have an alkali resistant technopolymer housing and accept different kinds of test tubes: straight tubes (100, 250, 400 ml and 1 L) or Kjeldahl balloon (500 ml).
Certificaten: Conformity with multiple standards protocols AOAC, ISO, EPA, DIN etc.
Informatie over toebehoren: Optional accessories such as test tubes, test tube spacer, test tube connection, connection cables and IQ/OQ/PQ manuals are available on request.
Leveringsinformatie: Each unit comes ready to use and is supplied with 250 ml test tube, 250 ml collecting flask, pincer for test tubes, touch pen and protective film for touch screen.