Anti-ABP1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BOSSBS-4733R-A647EA 437 EUR
BOSSBS-4733R-A647
Anti-ABP1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
ABP1

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR