Preparative HPLC system, Prep LC Premier
Leverancier: Knauer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Prep LC Premier
905-0671EA 306 EUR
905-0671 905-0670 KNAUA05852 KNAUA9850 905-0639 905-0638 905-0637 905-0636 905-0679 905-0635 905-0634 905-0678 905-0677 905-0556 905-0633 905-0676 905-0632 905-0554 KNAUCPE20KA38 905-0631 905-0675 905-0630 905-0674 905-0673 905-0672 905-0681 905-0680 905-0609 905-0607 905-0605 905-0648 905-0604 905-0603 905-0602 KNAUA9849-1 905-0601 905-0689 905-0688 905-0687 905-0686 KNAUA1033-2 905-0685 905-0684 KNAUA1033-1 KNAUCPE20LA38 905-0683 905-0693 905-0692 905-0691 905-0690 KNAUA05852-1 905-0619 905-0618 905-0617 905-0616 905-0615 905-0614 905-0613 905-0612 905-0655 905-0611 905-0654 905-0653 905-0652 905-0696 905-0651 KNAUCPE20MA38 905-0695 905-0650 905-0694 KNAUCPE20NA38 905-0629 905-0628 905-0627 905-0626 905-0625 905-0668 905-0624 905-0667 905-0623 905-0622 905-0666 905-0621 905-0665 905-0620
Preparative HPLC system, Prep LC Premier
Chromatografiesystemen Complete preparative HPLC-systemen
Prep LC Premier is a HPLC system for preparative applications, which are available as isocratic or gradient systems. The desired system is assembled from the Prep LC Premier elements that include pumps, detectors, and assistants. The assistants come equipped with up to three units, which are incorporated micro devices and offer many possibilities.
  • Designed for flexibility and to comfortably handle large sample volumes
  • Can be optimally adapted to the scale you need
The isocratic system consists of a pump P 2.1L, a detector UVD 2.1L, and an assistant ASM 2.1L. For a low pressure gradient system, you additionally need a valve block. For a high pressure gradient system, you need an additional pump P 2.1L.
The preparative HPLC system can be operated with chromatography software or Control Unit CU 2.1. The Control Unit is a touch display for direct control and optionally available. When it is connected to a device, it automatically recognises the device and provides the appropriate user interface. Note that you can control only one device at a time.
Prep LC Premier Compact Prep LC: This system fits on any lab bench and allows purification tasks up to 50 ml/min. Additional devices can be easily added any time due to the modular design of Prep LC Premier.
Prep LC Premier Lab Prep LC: The Lab Prep LC system is designed for your more demanding semi-preparative separations. You can customise a highly flexible LC system with the freely combinable components. With a maximum flow rate of 50 ml/min it is possible to separate up to several hundred milligrams per run.
Prep LC Premier Pilot Prep LC: Choose the Pilot Prep LC system if you want to increase your productivity even more. As for the Prep LC Premier Lab Prep LC system you can freely build up your system. Flow rates up to 1000 ml/min and loads up to several grams are possible. Optional peak and solvent recycling can be set up to increase separation power and reduce separation costs significantly.