Biological magnetic stirrer, multi-position, Techne® MCS-104S
Leverancier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Techne®
TECNFMCS104XEA 3499.87 EUR
TECNFMCS104X TECNFMCS104L TECNFMCS102L 442-0707
Biological magnetic stirrer, multi-position, Techne® MCS-104S
Roerders Magnetische roerders, meerdere posities
Multi-position magnetic stirrer designed for optimal suspension cell culture and the use of microcarriers. The system from Techne® consists of a stainless steel stirrer platform with locators for the glass culture vessels (not included). The design of the vessels, together with the bulb-ended stirrer, ensures that the cells are lifted into suspension at the lowest possible speeds. This gentle stirring action promotes high cell yields by preventing cell shearing.
  • Can accommodate up to four culture vessels simultaneously
  • Cell damage is avoided with soft start/stop for slow acceleration or deceleration (20 seconds)
  • Cell attachment and high yields promoted by interval setting option (6 s to 5 min on/2 min to 2 h off)
The soft start/stop ensures slow acceleration and deceleration of the stirrer, avoiding excessive turbulence in the culture media and eliminating cell damage. The interval stirring can be used during the attachment phase to further reduce agitation of the media or when culturing particularly fragile cells. The stirrers create virtually no heat so there is negligible heat transfer from the magnetic drive to the culture vessel, making the system suitable for use in incubators and cold rooms up to 40 °C and 95% relative humidity (non-condensing).
Bestelinformatie: Culture vessels must be ordered separately. Other stirrer models accommodating different sizes and numbers of vessels are available. Culture vessels from 125 ml to 5 litres are available, please contact your local Avantor office for details.