Procedure mask, type II
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ANSM85001EA 6.6 EUR
ANSM85001
Procedure mask, type II
Maskers Hygiëne maskers
Procedure mask helps prevent large particles expelled by the wearer such as spit or mucous from reaching the patient or work environment.
  • Pleat style with ear loops
  • Protective three-layer construction
Appropriate for situations where exposure to blood and other body fluids from the patient or work environment.