Zoutzuur 0.5 N, Biotech Reagent, J.T.Baker®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Filtered through a 0.2 µm filter.
0335-07EA
0
EUR
0335-07
Zoutzuur 0.5 N, Biotech Reagent, J.T.Baker®
Zoutzuur
Made from USP purified water and hydrochloric acid, NF (multi-compendial) which meets BP, Ph. Eur. and JP chemical specifications.
Formule:
HCl MW: 36,46 g/mol Dichtheid: 1,00 g/cm³ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011324 CAS nummer: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Specification Test Results
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Appearance of Solution | Passes Test |
Identification A | Passes Test |
Identification B | Passes Test |
Normality | 0.485 - 0.515 |
Residue on Ignition | ≤ 0.008 % |
Ammonium (NH₄) | ≤ 3 ppm |
Trace Impurities - Heavy Metals (as Pb) | ≤ 0.5 ppm |
Trace Impurities - Iron (Fe) | ≤ 0.10 ppm |
Sulfate (SO₄) | ≤ 1 ppm |
Trace Impurities - Arsenic (As) | ≤ 2.000 ppm |
Made from U.S.P. Purified Water and Hydrochloric Acid, N.F. (Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications | |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. | |
Filtered through a 0.2 micron filter. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...