Laboratory blenders
Leverancier: Waring
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WARICB15EEA 4210 EUR
WARICB15E WARICB15TE
Laboratory blenders
Homogenisatoren Standaard laboratoriumblenders
The CB15 series blenders are designed specifically for processing large quantities of free flowing material with a high liquid content. This range of 3-speed blenders has an integral pulse button and is fitted with a powerful 3,75 hp motor encased within a die-cast epoxy coated motor housing.
  • Easy to move and fitted with full traction rubber feet for greater stability
  • Easy-to-clean electronic keypad
  • Slow Start™ blending mechanism
  • Stainless steel container jar with two handles
  • Smaller 2, 1 litre and mini-containers can be used with the aid of the AD adapters
In addition the CB15VXE model has an a built in tachometer display showing the RPM of the motor. This makes it suitable for a range of validated applications such as GMP/GLP.