Dispenser unit for tissue embedding system, TES 99
Leverancier: Medite
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MEDT02-9925.00EA 0 EUR
MEDT02-9925.00
Dispenser unit for tissue embedding system, TES 99
Houders voor het inbedden van weefsel
The dispenser unit with its large working area and the heat protective hand rest provides comfortable embedding.
  • Connections for 2× forceps, heating for 6× forceps
Convenient for use with all kinds of systems and tissue specimens of any size.
Electrical information: 115 - 230 V; 50/60 Hz; 350 VA
Spot size Ø 40 mm at –5 °C