Netherlands
Bestelling invoeren
Avantor® Hichrom Presat, HPLC Columns
Avantor® Hichrom Presat, HPLC Columns
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Avantor® Hichrom Presat, HPLC Columns
Supplier:  Avantor
   Sustainable Options Available
Specifications
  • Catalog No:
  • HICH28610
  • HICH28650
  • Environmentally Preferable:
  • Lage impact op de fabricage
  • Lage impact op de fabricage
  • Milieu certificering:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
Specifications
Product Information
  • Catalog #
    Kolomtype
    Fase
    Kolombehuizing
    Ø inw.
    Lengte
    Verpakkingsmateriaal
    VPE
    Availability
    Price
  • HICH28610
    Analytisch
    Presaturation
    Roestvrij staal
    4,60 mm
    250 mm
    Silica
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Local Article Number: HICH28610
    Specifications:
      Lage impact op de fabricage
    More
    Specifications
    Environmentally Preferable
    Lage impact op de fabricage
    Milieu certificering
    ISO 14001:2015 (530840 UM15)
      Lage impact op de fabricage
  • HICH28650
    Analytisch
    Presaturation
    Roestvrij staal
    4,60 mm
    250 mm
    Silica
    4
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST
    Local Article Number: HICH28650
    Specifications:
      Lage impact op de fabricage
    More
    Specifications
    Environmentally Preferable
    Lage impact op de fabricage
    Milieu certificering
    ISO 14001:2015 (530840 UM15)
      Lage impact op de fabricage