VWR®, Weighing Bottles, Glass
Supplier: VWR Collection
Certificates
About this item
Borosilicate glass 3,3, low form.
- Interchangeable knobbed lid with ground joint
- White labelling area
- Resistant to chemicals and thermal shock
Related Information
Product Information
- Catalog #CapacityØ×HNSPkAvailabilityPrice
- Local Article Number: VWRI611-3817Specifications:
Specifications
Environmentally Preferable - Local Article Number: VWRI611-3818Specifications:
Specifications
Environmentally Preferable - Local Article Number: VWRI611-3819Specifications:
Specifications
Environmentally Preferable - Local Article Number: VWRI611-3820Specifications:
Specifications
Environmentally Preferable - Local Article Number: VWRI611-3821Specifications:
Specifications
Environmentally Preferable - Local Article Number: VWRI611-3822Specifications:
Specifications
Environmentally Preferable