Hitachi LC-besturingsoptie voor Empower 2
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR® Hitachi
906-0090EA 0 EUR
906-0090 906-0091 HITA892-0012S HITA892-0011S 906-0156 906-0158 906-0159 906-0160
Hitachi LC-besturingsoptie voor Empower 2
Software Laboratoriumsoftware
Empower 3 is Waters's compliant-ready chromatography data software (CDS) package for advanced data acquisition, management, processing, reporting and distribution. Empower 3's interface is designed to maximise laboratory efficiency, improving collection, processing and printing chromatography data. Capabilities include:
  • Customisable data reports
  • Integrated custom calculations
  • Relational data base so all the meta data is traceable back to the raw data
  • Optimised workflow for different users and different analyses
With enhanced 21 CFR Part 11 compliance capabilities, Empower 3 takes the laboratory's ability to manage data integrity, advanced security features and audit trails to new levels. Empower 3 can grow seamlessly from a single workstation to an enterprise-wide system with Empower 3's scalable, modular architecture, which fits into the most modern, secure, HA (high availability) corporate architecture.