Cool unit –15 °C for tissue embedding system, TES 99
Leverancier: Medite
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MEDT02-9941-00EA 3150 EUR
MEDT02-9941-00
Cool unit –15 °C for tissue embedding system, TES 99
Koelers Tafelmodel koelers
This cold plate with a powerful aggregate easily reaches a temperature of –15 °C in very short time.
Its working surface can accomodate up to 110 blocks.