Bioreactors, UniVessel® SU
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SARPDUO002LL-SS01V
Bioreactors, UniVessel® SU
Bioreactoren
The UniVessel® SU is a stirred tank, single-use bioreactor with a working volume range of 0,6 to 2 L. It is assembled, irradiated and shipped ready-to-use.
  • Proven and scaleable design
  • Compatible with your existing bioreactor controller
  • Interchangeable with existing glass vessels
UniVessel® SU is compatible with your controller and can be used interchangeably with glass vessels to help you efficiently manage peak workloads despite challenging timelines.