Low alloy steel standard
Leverancier: LGC Standards
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PROONIST-1764AEA 0 EUR
PROONIST-1764A
Low alloy steel standard
Standaarden Legeringen en metaalstandaarden
This reference material is intended for use in the evaluation of chemical and instrumental analysis and in calibration of those methods.
  • 22 analytes
A unit consists of a disk approximately 34 mm in diameter and 19 mm thick.
Certificaten: Traceable to NIST and SRM.