Lithium ore, spodumene - lithium oxide, NIST standards
Leverancier: LGC Standards
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PROONIST-182EA 0 EUR
PROONIST-182
Lithium ore, spodumene - lithium oxide, NIST standards
Standaarden Legeringen en metaalstandaarden
This reference standards are intended to use in checking the accuracy of assay methods. These are supplied in fine powder form.
Certificaten: Traceable to NIST and SRM numbers.