Combustion analyser, 310
Leverancier: Testo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TEST0563.3100EA 384 EUR
TEST0563.3100
Combustion analyser, 310
The 310 residential combustion analyser provides value and expanded capabilities in combustion testing with its rugged housing, simple design and advanced sensor technology.
  • Start testing faster in only 30 seconds
  • Easy-to-read backlit display shows more measurements which provides a more complete picture of the combustion process
  • Simple top-of-display icons show you the current measurement functions
  • Kink-resistant hose is integrated into the housing to eliminate leaks
  • Stainless steel probe and cone-stop is perfect for residential furnace, boiler and water heater applications
Five different fuels to select from (natural gas, propane, fuel oil, biomass 5%, wood 20 %), it provides the flexibility needed for all tuning opportunities.
Fully featured for better testing and tuning of undiluted CO (CO air free), ambient CO, draft and pressure, temperature (flue and ambient), combustion/flue gas efficiency and excess air.
Bestelinformatie: Includes analyser, battery, sensors, probe with cone, hose, silicone tubing (for pressure), extra particulate filters (5), AC power supply (USB), pressure plugs (5), calibration certificate and case.