Netherlands
Order Entry
Accessories for container PACKO
Supplier:  CURTEC
Certificates
undefined
Accessories for container PACKO
 :  CURTEC
Product Information
  • Catalog #
    Supplier #
    Cap colour
    Pk
    Availability
    Price
  • Screw lid, for 300 - 1300 ml packo container
    Screw lid, for 300 - 1300 ml packo container
    CURT4350-70-402
    4350-70-402
    Red
    210
    Log in for availability
    1 * 210 ST
    1 * 210 ST - 195,00 €
    195,00 €
    1 * 210 ST
       195,00 €
    Supplier Article Number: 4350-70-402
    Local Article Number: CURT4350-70-402
    Specifications
    Environmentally Preferable
  • Screw lid, for 300 - 1300 ml packo container
    Screw lid, for 300 - 1300 ml packo container
    CURT4350-70-480
    4350-70-480
    Dark red
    210
    Log in for availability
    1 * 210 ST
    1 * 210 ST - 218,00 €
    218,00 €
    1 * 210 ST
       218,00 €
    Supplier Article Number: 4350-70-480
    Local Article Number: CURT4350-70-480
    Specifications
    Environmentally Preferable
  • Screw lid, for 300 - 1300 ml packo container
    Screw lid, for 300 - 1300 ml packo container
    CURT4350-70-505
    4350-70-505
    Green
    210
    Log in for availability
    1 * 210 ST
    1 * 210 ST - 178,00 €
    178,00 €
    1 * 210 ST
       178,00 €
    Supplier Article Number: 4350-70-505
    Local Article Number: CURT4350-70-505
    Specifications
    Environmentally Preferable