Ferromanganese, high carbon, alloy, NIST-68C, reference standard
Leverancier: LGC Standards
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PROONIST-68CEA 0 EUR
PROONIST-68C
Ferromanganese, high carbon, alloy, NIST-68C, reference standard
Standaarden Legeringen en metaalstandaarden
This standard reference material is in the powder form intended for use in checking chemical methods of analysis and in calibration with instrumental methods of analysis.
  • 8 analytes/elements measured
Certificaten: Traceable to NIST and SRM.