Human IgG1 Isotype Control
Catalog # PRSI96-306
Supplier: ProSci Inc.
- Antibody type:Control
- Host:Human
- Isotype:IgG1
- Form:Lyophilized
- Purity:>95% as determined by SDS-PAGE.
- Storage buffer:50 mM tris, 100 mM glycine, pH7.5
- Molecular weight:26 kDa
- Tested applications:Western Blot
- Size:0.2 mg
- Pk:200 µG
About this item
Crystallizable fragments composed of the carboxy-terminal halves of both IMMUNOGLOBULIN HEAVY CHAINS linked to each other by disulfide bonds. Fc fragments contain the carboxy-terminal parts of the heavy chain constant regions that are responsible for the effector functions of an immunoglobulin (COMPLEMENT fixation, binding to the cell membrane via FC RECEPTORS, and placental transport). IgG1 Fc was reported has a novel role as a potential anti-inflammatory drug for treatment of human autoimmune diseases.