Netherlands
Order Entry
Hafnium standard solution, for ICP, in 5% HCl, VHG
Hafnium standard solution, for ICP, in 5% HCl, VHG
Certificates
Hafnium standard solution, for ICP, in 5% HCl, VHG
Certificates
Related Information
Product Information
  • Catalog #
    Supplier #
    Element
    Symbol
    Concentration
    Matrix
    Pk
    Availability
    Price
  • PROOVHG-PHFH-100
    VHG-PHFH-100
    Hafnium
    Hf
    1000 mg/L
    5% HCl
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL
  • PROOVHG-PHFH-500
    VHG-PHFH-500
    Hafnium
    Hf
    1000 mg/L
    5% HCl
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL
  • PROOVHG-THFH-100
    VHG-THFH-100
    Hafnium
    Hf
    10000 mg/L
    5% HCl
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL
  • PROOVHG-THFH-500
    VHG-THFH-500
    Hafnium
    Hf
    10000 mg/L
    5% HCl
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL
This product has been blocked by your organization. Please contact your purchasing department for more information
Frequently Bought Together