Neodymium standard solution, for ICP, in 5% HNO₃, VHG
Supplier: LGC Standards PROMOCHEM
Certificates
About this item
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.
Product Information
- Catalog #Supplier #ElementSymbolConcentrationMatrixPk InfoPkAvailabilityPrice
- PROOVHG-PNDN-500VHG-PNDN-500NeodymiumNd1000 μg/ml5% HNO₃500 ml500 mLLog in for availability1 * 500 mL
- PROOVHG-TNDN-100VHG-TNDN-100NeodymiumNd10000 μg/ml5% HNO₃100 ml100 mLLog in for availability1 * 100 mL
- PROOVHG-TNDN-500VHG-TNDN-500NeodymiumNd10000 μg/ml5% HNO₃500 ml500 mLLog in for availability1 * 500 mL
This product has been blocked by your organization. Please contact your purchasing department for more information
Frequently Bought Together