RNP transfection reagent for genome editing, jetCRISPR™
Leverancier: Polyplus-transfection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
502-07EA 334 EUR
PPLU502-07 PPLU502-01
RNP transfection reagent for genome editing, jetCRISPR™
jetCRISPR™ is a RNP delivery reagent, specially designed for co-delivery of guide RNA and cas9 protein for gene editing experiments.
  • Excellent cell viability and morphology throughout the whole duration of the experiments
  • Fast and reliable gene editing
  • Easy to use: reverse and forward protocols
The CRISPR/Cas9 engineered nuclease system is a powerful and highly specific genome editing tool. It is provided as a ready to use solution specifically designed for high transfection efficiency of RNP, thus leading to a high cas9-mediated genome editing efficiency on a wide variety of targets in different cell types.
Certificaten: Polyplus-transfection® has been an ISO 9001 accredited company since 2002. Since then this certification has been successfully renewed every three years.
Bestelinformatie: 1,5 ml of jetCRISPR™ transfection reagent is sufficient to perform up to ~1250 transfections in 24-well plates or ~300 transfections in 6-well plates. Bulk quantities are also available upon request, please contact your local Avantor sales office for more details.