Human mononuclear cells from cord blood
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HCMSCB009C-1EA 0 EUR
HCMSCB009C-1 HCMSCB009C-2
Human mononuclear cells from cord blood
Mononuclear cells are critical components of the immune system which are involved in both humoral and cell-mediated immunity. The mononuclear cell is a widely used product in research and clinical applications such as in microbiology, virology, oncology, vaccine development, transplant and regenerative biology, and toxicology.
  • Tested negative for HIV, HTLVI/II, HCV, HBV, WNV, syphilis, CMV, NAT, NAT WNV and Chagas
  • Cryopreserved in CryoStor® CS10 media
Human cord blood is collected aseptically at the time of delivery from healthy pregnant women under IRB approved informed consent. Mononuclear cells are isolated by diluting the cord blood with balanced saline and using density gradient centrifugation techniques. It can be further processed to isolate a variety of sub populations.
Storage: Either prepare cells for long term storage in liquid nitrogen vapour phase or thaw for use. Storage in liquid phase nitrogen is NOT recommended. Short term storage of cells (less than 2 weeks) at –80 °C is acceptable, but should be minimised to ensure maximum stability. Once thawed, samples must be used immediately.
Opgelet: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.