Whole blood, multiple myeloma
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HCMSPB000F-2-MMEA 0 EUR
HCMSPB000F-2-MM
Whole blood, multiple myeloma
Whole blood used for researching the causes, diagnosis, supportive care and treatment of Lung Cancer (LC). Patient samples can be used by researchers to investigate biomarker and diagnostic development, pre-clinical research validation, and drug toxicity and efficacy. Collected from IRB-consented donors diagnosed with multiple myeloma.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV Nucleic Acid, WNV Nucleic Acid, Trypanasoma cruzi and Syphilis.
Storage: Use immediately upon receipt.
Opgelet: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.