Whole blood, chronic lymphocytic leukemia (CLL)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HCMSPB000F-2-CLLEA 0 EUR
HCMSPB000F-2-CLL
Whole blood, chronic lymphocytic leukemia (CLL)
Whole blood used for researching the causes, diagnosis, supportive care and treatment of chronic lymphocitic leukemia (CLL). The whole blood is collected from IRB-consented donors diagnosed with CLL.
No pre screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV Nucleic Acid, WNV Nucleic Acid, Trypanasoma cruzi and Syphilis.
Storage: Use immediately upon receipt.
Opgelet: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.