Whole blood, herpes simplex virus (HSV)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HCMSPB000F-2-HSV2EA 0 EUR
HCMSPB000F-2-HSV2 HCMSPB000F-2-HSV1
Whole blood, herpes simplex virus (HSV)
Whole blood used for researching the causes, diagnosis, supportive care and treatment of herpes simplex virus 1 and 2 (HSV 1 and HSV 2). Whole blood is collected from IRB-consented donors diagnosed with herpes simplex virus (HSV).
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV Nucleic Acid, WNV Nucleic Acid, Trypanasoma cruzi and Syphilis.
Storage: Use immediately upon receipt.
Opgelet: Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.