Formate ion chromatography standards, VHG
Leverancier: LGC Standards
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VHG
PROOVHG-IFORM-500EA 0 EUR
PROOVHG-IFORM-500 PROOVHG-IFORM-100
Formate ion chromatography standards, VHG
Standaarden Anionische standaarden Formiaat standaarden
This solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Certificaten: Traceable to NIST and CRM.