Engraving device with points
Leverancier: Bochem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BOCH12601EA 139 EUR
BOCH12601
Engraving device with points
This engraving device is used for permanent labelling of lab equipment.
  • Engraves metal, glass and plastic
  • Engraving through vibration principle
Leveringsinformatie: Engraving device in practical carrying case includes four points.