Alcohol-based rapid disinfectant, ready to use, deconex® SOLARSEPT
Leverancier: Binder
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIND1002-0022EA 109 EUR
BIND1002-0022
Alcohol-based rapid disinfectant, ready to use, deconex® SOLARSEPT
Reinigingsmiddelen Desinfectiemiddelen
deconex® SOLARSEPT is used for disinfection when a short exposure and drying time is required.
  • aldehyde-free
  • Ready-to-use, with rapid effect ime and short drying time
  • Can also be sprayed on
  • Broad spectrum of activity including tuberculocidal effect
  • Pleasant fresh smell
Binder recommend alcohol based solutions for exterior surfaces, inner chamber, racks and door gaskets.
Wipe disinfection: Pour the product out of the dosing bottle onto a clean cloth or directly onto the surface to be treated (approx. 25 ml product/m²).
Gently wipe the surface all over with steady movements. Makesure that the surface is completely wetted.
Spray disinfection: Spray the surface to be treated, ensuring in particular that it is completely wetted.
Certificaten: Certified by VAH (German Association for Applied Hygiene)
Opgelet: Due to the alcohol content, spraying is limited for safety reasons to small areas which cannot be reached by wiping.