Photoionisation detector with PPC add-on kit
Leverancier: Perkin Elmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PERCN6120131EA 19450 EUR
PERCN6120131
Photoionisation detector with PPC add-on kit
Chromatografie detectoren
Photoionisation detector with programmable pneumatic control (PPC) add-on kit for gas chromatography.
Leveringsinformatie: Includes a detector, heater, sensor, heater block, and programmable pneumatic control for make-up gas.