Netherlands
Order Entry
Barrels with lid and clamping ring
Barrels with lid and clamping ring
 :  RIXIUS
undefined
Barrels with lid and clamping ring
 :  RIXIUS
  •  
    Capacity
    Ext.Ø
    Ø opening
    Height
    Pk
  • UN approval: 1H2/X60/S
    UN approval: 1H2/X60/S
    216-5378
    5-2103-030-GM
    30 L
    340 mm
    251 mm
    460 mm
    1
    1 * 1 ST
    1 * 1 ST - 55,90 €
    55,90 €
    1 * 1 ST
       55,90 €
     : 5-2103-030-GM
     : RIXI5-2103-030-GM
    Accessories
    Specifications
    Environmentally Preferable
  • UN approval: 1H2/X88/S
    UN approval: 1H2/X88/S
    216-1627
    5-2103-060-MA
    60 L
    400 mm
    325 mm
    610 mm
    1
    1 * 1 ST
    1 * 1 ST - 86,40 €
    86,40 €
    1 * 1 ST
       86,40 €
     : 5-2103-060-MA
     : RIXI5-2103-060-MA
    Accessories
    Specifications
    Environmentally Preferable
  • UN approval: UN 1H2/X225/S
    UN approval: UN 1H2/X225/S
    216-1628
    5-2103-120-MA
    120 L
    500 mm
    396 mm
    800 mm
    1
    1 * 1 ST
    1 * 1 ST - 143,00 €
    143,00 €
    1 * 1 ST
       143,00 €
     : 5-2103-120-MA
     : RIXI5-2103-120-MA
    Accessories
    Specifications
    Environmentally Preferable
  • UN approval: 1H2/X278/S/D/BAM 8281
    UN approval: 1H2/X278/S/D/BAM 8281
    RIXI5-2103-150-MA
    5-2103-150-MA
    150 L
    — mm
    — mm
    970 mm
    1
    1 * 1 ST
    1 * 1 ST - 157,00 €
    157,00 €
    1 * 1 ST
       157,00 €
     : 5-2103-150-MA
     : RIXI5-2103-150-MA
    Accessories
    Specifications
    Environmentally Preferable
  • UN approval: 1H2/X318/S
    UN approval: 1H2/X318/S
    216-5660
    5-2103-220-MA
    220 L
    590 mm
    460 mm
    970 mm
    1
    1 * 1 ST
    1 * 1 ST - 249,00 €
    249,00 €
    1 * 1 ST
       249,00 €
     : 5-2103-220-MA
     : RIXI5-2103-220-MA
    Accessories
    Specifications
    Environmentally Preferable