Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Netherlands
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Titanium standard solution, for ICP, in H₂O, tr. F⁻ , CRM, VHG
:
LGC Standards PROMOCHEM
Titanium standard solution, for ICP, in H₂O, tr. F⁻ , CRM, VHG
:
LGC Standards PROMOCHEM
Element
Symbol
Concentration
Matrix
Pack type
Pk
PROOVHG-PTIW-100
VHG-PTIW-100
Titanium
Ti
1000 μg/ml
H₂O/tr HF
100 ml
100 mL
1 * 100 mL
-
+
1 * 100 mL
-
+
PROOVHG-PTIW-500
VHG-PTIW-500
Titanium
Ti
1000 μg/ml
H₂O/tr HF
500 ml
500 mL
1 * 500 mL
-
+
1 * 500 mL
-
+
PROOVHG-TTIW-100
VHG-TTIW-100
Titanium
Ti
10000 μg/ml
H₂O/tr HF
100 ml
100 mL
1 * 100 mL
-
+
1 * 100 mL
-
+
PROOVHG-TTIW-500
VHG-TTIW-500
Titanium
Ti
10000 μg/ml
H₂O/tr HF
500 ml
500 mL
1 * 500 mL
-
+
1 * 500 mL
-
+