Tungsten standard solution, for ICP, in 5% HNO₃, tr. HF, CRM, VHG
Supplier: LGC Standards PROMOCHEM
Certificates
About this item
Product Information
- Catalog #Supplier #ElementSymbolConcentrationMatrixPkAvailabilityPrice
- PROOVHG-PWNF-100VHG-PWNF-100TungstenW1000 μg/ml5% HNO₃100 mL1 * 100 mLSupplier Article Number: VHG-PWNF-100Local Article Number: PROOVHG-PWNF-100
- PROOVHG-PWNF-500VHG-PWNF-500TungstenW1000 μg/ml5% HNO₃500 mL1 * 500 mLSupplier Article Number: VHG-PWNF-500Local Article Number: PROOVHG-PWNF-500
- PROOVHG-TWNF-100VHG-TWNF-100TungstenW10000 μg/ml5% HNO₃100 mL1 * 100 mLSupplier Article Number: VHG-TWNF-100Local Article Number: PROOVHG-TWNF-100
- PROOVHG-TWNF-500VHG-TWNF-500TungstenW10000 μg/ml5% HNO₃500 mL1 * 500 mLSupplier Article Number: VHG-TWNF-500Local Article Number: PROOVHG-TWNF-500