Netherlands
Order Entry
VWR® Rotors for Ministar Blueline Centrifuge
VWR® Rotors for Ministar Blueline Centrifuge
Supplier:  VWR Collection
Certificates
VWR® Rotors for Ministar Blueline Centrifuge
Supplier:  VWR Collection
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Replacement tube rotor, quick release, 8×1,5/2,0 ml tubes, for MiniStar blueline centrifuge
    Replacement tube rotor, quick release, 8×1,5/2,0 ml tubes, for MiniStar blueline centrifuge
    521-2323
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 74,10 €
    74,10 €
    1 * 1 ST
       74,10 €
    Local Article Number: VWRI521-2323
    Specifications:
    Specifications
    Environmentally Preferable
  • Replacement strip tube rotor, quick release, 4× (8×0,2 ml) PCR strip tubes, for MiniStar blueline centrifuge
    Replacement strip tube rotor, quick release, 4× (8×0,2 ml) PCR strip tubes, for MiniStar blueline centrifuge
    521-2322
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 75,80 €
    75,80 €
    1 * 1 ST
       75,80 €
    Local Article Number: VWRI521-2322
    Specifications:
    Specifications
    Environmentally Preferable