VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
Certificates
About this item
Cardboard, coated.
- Waterproof coating
- Freezable to –140 °C
- Available with or without dividers
- In different heights and five colours
For storage of cryovials at low temperatures.
Related Information
Boxes, without dividers
Product Information
- Catalog #CompartmentsColourW×D×H ext.PkAvailabilityPrice
- Local Article Number: VWRI479-7020Specifications:
Specifications
Environmentally Preferable - Local Article Number: VWRI479-7021Specifications:
Specifications
Environmentally Preferable - Local Article Number: VWRI479-7019Specifications:
Specifications
Environmentally Preferable - Local Article Number: VWRI479-7022Specifications:
Specifications
Environmentally Preferable - Local Article Number: VWRI479-7023Specifications:
Specifications
Environmentally Preferable - Local Article Number: VWRI479-7024Specifications:
Specifications
Environmentally Preferable - Local Article Number: VWRI479-7025Specifications:
Specifications
Environmentally Preferable
Boxes with dividers
Product Information
- Catalog #CompartmentsColourW×D×H ext.PkAvailabilityPrice
- Local Article Number: VWRI479-7031Specifications:
Specifications
Environmentally Preferable - Local Article Number: VWRI479-7028Specifications:
Specifications
Environmentally Preferable - Local Article Number: VWRI479-7032Specifications:
Specifications
Environmentally Preferable - Local Article Number: VWRI479-7029Specifications:
Specifications
Environmentally Preferable - Local Article Number: VWRI479-7026Specifications:
Specifications
Environmentally Preferable - Local Article Number: VWRI479-7027Specifications:
Specifications
Environmentally Preferable - Local Article Number: VWRI479-7030Specifications:
Specifications
Environmentally Preferable
Dividers for cryoboxes
Product Information
- Catalog #CompartmentsColourW×D×H ext.PkAvailabilityPrice
- For cryoboxes 95 mm heightLocal Article Number: VWRI479-7035Specifications:
Specifications
Environmentally Preferable - For cryoboxes 55 mm heightLocal Article Number: VWRI479-7033Specifications:
Specifications
Environmentally Preferable - For cryoboxes 75 mm heightLocal Article Number: VWRI479-7034Specifications:
Specifications
Environmentally Preferable