Netherlands
Order Entry
Industrial gloves, ActivArmr® 97-631
Industrial gloves, ActivArmr® 97-631
 :  Ansell
undefined
Industrial gloves, ActivArmr® 97-631
 :  Ansell
  • Catalog No:
  • ANSE97-631/7
  • ANSE97-631/9
  • ANSE97-631/10
  • ANSE97-631/11
  • ANSE97-631/8
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Nylon/acrylic
  • Nylon/acrylic
  • Nylon/acrylic
  • Nylon/acrylic
  • Nylon/acrylic
  • Finish:
  • PVC coating on palm
  • PVC coating on palm
  • PVC coating on palm
  • PVC coating on palm
  • PVC coating on palm
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (2231B), EN 511 (12X)
  • EN 388:2016 (2231B), EN 511 (12X)
  • EN 388:2016 (2231B), EN 511 (12X)
  • EN 388:2016 (2231B), EN 511 (12X)
  • EN 388:2016 (2231B), EN 511 (12X)
  • Food contact approved:
  • +
  • +
  • +
  • +
  • +
  •  
    Glove size
    Colour
    Length
    Packed
    Pk
  • ANSE97-631/7
    97-631/7
    7
    Black
    250 - 285 mm
    6 pais/bag
    6 PAIR
    1 * 6 PAAR
    1 * 6 PAAR - 70,50 €
    70,50 €
    1 * 6 PAAR
       70,50 €
     : 97-631/7
     : ANSE97-631/7
    Specifications
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    Nylon/acrylic
    Finish
    PVC coating on palm
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (2231B), EN 511 (12X)
    Food contact approved
    +
  • ANSE97-631/8
    97-631/8
    8
    Black
    250 - 285 mm
    6 pais/bag
    6 PAIR
    1 * 6 PAAR
    1 * 6 PAAR - 70,50 €
    70,50 €
    1 * 6 PAAR
       70,50 €
     : 97-631/8
     : ANSE97-631/8
    Specifications
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    Nylon/acrylic
    Finish
    PVC coating on palm
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (2231B), EN 511 (12X)
    Food contact approved
    +
  • ANSE97-631/9
    97-631/9
    9
    Black
    250 - 285 mm
    6 pais/bag
    6 PAIR
    1 * 6 PAAR
    1 * 6 PAAR - 70,50 €
    70,50 €
    1 * 6 PAAR
       70,50 €
     : 97-631/9
     : ANSE97-631/9
    Specifications
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    Nylon/acrylic
    Finish
    PVC coating on palm
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (2231B), EN 511 (12X)
    Food contact approved
    +
  • ANSE97-631/10
    97-631/10
    10
    Black
    250 - 285 mm
    6 pais/bag
    6 PAIR
    1 * 6 PAAR
    1 * 6 PAAR - 70,50 €
    70,50 €
    1 * 6 PAAR
       70,50 €
     : 97-631/10
     : ANSE97-631/10
    Specifications
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    Nylon/acrylic
    Finish
    PVC coating on palm
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (2231B), EN 511 (12X)
    Food contact approved
    +
  • ANSE97-631/11
    97-631/11
    11
    Black
    250 - 285 mm
    6 pais/bag
    6 PAIR
    1 * 6 PAAR
    1 * 6 PAAR - 70,50 €
    70,50 €
    1 * 6 PAAR
       70,50 €
     : 97-631/11
     : ANSE97-631/11
    Specifications
    Ambidextrous / hand-specific
    Hand-specific
    Environmentally Preferable
    Disposable / reusable
    Reusable
    Liner material
    Nylon/acrylic
    Finish
    PVC coating on palm
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (2231B), EN 511 (12X)
    Food contact approved
    +