Anti-NF-kB p65 Polyclonal Antibody
Catalog # BSBTPB9324
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:NF-kB p65
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for NFkB p65/RELA detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Mouse. NFkB p65/RELA information: Molecular Weight: 60212 MW; Subcellular Localization: Nucleus . Cytoplasm . Colocalized with DDX1 in the nucleus upon TNF-alpha induction (By similarity). Nuclear, but also found in the cytoplasm in an inactive form complexed to an inhibitor (I-kappa-B). Colocalizes with GFI1 in the nucleus after lipopolysaccharide (LPS) stimulation.
Type: Primary
Antigen: NF-kB p65
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse; Rat