Avantor® Hichrom Genesis, HPLC Columns, C4-300
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HICH8P25976EA 2210 EUR
HICH8P25976
Avantor® Hichrom Genesis, HPLC Columns, C4-300
Avantor® Genesis phases are based on high purity metal-free spherical silica. They are suitable for the analysis of a wide range of analytes.
  • Good peak shape and reproducibility
  • Long column lifetime
  • pH stability 1 to 10