Bioassay dishes, Nunc™
Supplier: Thermo Fisher Scientific
Certificates
PS, sterile, with lid. Useful for screening large numbers of colonies in cloning experiments. Low profile dishes designed for genomic screening and colony picking.
- Can be used for culturing bacteria and fungi, also suitable in agar diffusion assays
- Can be used as a moisture chamber for incubation of MicroWell™ plates
- Lined with wet filter paper can also be used as an incubation chamber for in situ hybridisation
- Large growth area
- Low profile dish fits robotic instruments
Specifications
- Catalog No:
- 734-2179
- 734-1342
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
Specifications
Product Information
- Catalog #Supplier #Recommended working volumeGrowth areaW×D×HPkAvailabilityPrice
- Bioassay dishes, PS, with lidBioassay dishes, PS, with lid734-2179240835225 ml500 cm²245×245×25 mm161 * 16 ST215,00 €
Specifications
Environmentally PreferableEnvironmental certification - Bioassay dishes, PS, with lid, low profileBioassay dishes, PS, with lid, low profile734-1342240845215 ml478 cm²241×241×20 mm201 * 20 ST305,00 €
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact