ESD occupational shoes, clogs, HACCP, Arrow, 38320
Leverancier: Abeba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Arrow
ABEB38320/40EA 57.8 EUR
ABEB38320/40 ABEB38320/41 ABEB38320/42 ABEB38320/43 ABEB38320/44 ABEB38320/45 ABEB38320/35 ABEB38320/46 ABEB38320/47 ABEB38320/36 ABEB38320/37 ABEB38320/38 ABEB38320/39
ESD occupational shoes, clogs, HACCP, Arrow, 38320
ESD clogs made from perforated microfibre upper, microfibre lining and a foldable and adjustable heel strap.
  • Sanitized® treatment reduces odour and bacterial growth
  • Abrasion and slip resistant PU sole
  • Washable at 30 °C
Colour: White
Certificaten: EN 61340 (ESD); HACCP; EN ISO 20347:2012; OB, A, FO, SRA