Flow indicator
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GRSGWR1-025GMWEA 313 EUR
GRSGWR1-025GMW
Flow indicator
Mechanical flow indicator for fluid media with rotor for quantitative display.
  • Robust construction in brass
  • Internal wiper to clean the glass without removing the device
  • Rotor visible from 360°
The sliding bearing of this flow indicator is made from PEEK. Brass construction includes CW614N nickelled, borosilicate glass, NBR.