Geleidbaarheidscellen, InLab® ISM®
Leverancier: Mettler-Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ISM®
663-0270EA 830 EUR
663-0270 663-0269 663-0268 663-0145 663-0178 663-0177 663-0000 663-0176
Geleidbaarheidscellen, InLab® ISM®
Elektroden Geleidbaarheidselektroden
High performance sensors combining robustness with state-of-the-art measurement technology. Compatible with either SevenExcellence™ and SevenCompact™ meters (663-0176/663-0177/663-0178) or with SevenGo Duo™ SG23 and SevenGo Duo pro™ SG78 meters (663-000/663-0145). Fully traceable.
The revolutionary intelligent sensor management system (ISM®) offers the user an even greater level of security. The calibration data is stored on a chip in the sensor and is used automatically when the sensor is connected to the meter. This ensures that the meter always uses the latest calibration data.
Temperature range: 0 to +100 °C (with NTC 30 kΩ)
Bestelinformatie: Supplied with quality certificate.